Dichloromethane, 99.8%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
Dichloromethane, 99.8%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
Dichloromethane, 99.8%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
Dichloromethane, 99.8%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
Thermo Scientific Chemicals
Dichloromethane, 99.8%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™
Dichloromethane, 99.8%, CH2Cl2, CAS Number-75-09-2, 513-35-9, aerothene mm, freon 30, methane dichloride, methane, dichloro, methylene bichloride, methylene chloride, methylene dichloride, narkotil, solaesthin, solmethine, 1L, 01, 60, 39.0 deg.C to 40.0 deg.C, CHEBI:15767, Colorless, >120 deg.C | CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA348460025
Price (MYR)
868.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
868.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDichloromethane
CAS75-09-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing cancer.
May cause drowsiness or dizziness.
CAS513-35-9