4-Methylmorpholine N-oxide, 50 wt.% solution in water
CAS: 7529-22-8 | C5H11NO2 | 117.15 g/mol
Have Questions?
Change view
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA348271000
Price (MYR)
321.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
321.00
EA
Add to cart
Chemical Identifiers
CAS7529-22-8, 7732-18-5
Specifications
Chemical Name or Material4-Methylmorpholine N-oxide
Name Note50 wt.% solution in water
CAS7529-22-8
CAS Min %50.0
Health Hazard 1GHS Signal Word: Warning