Dimethyltin dichloride, 95%
Dimethyltin dichloride, 95%
Dimethyltin dichloride, 95%
Dimethyltin dichloride, 95%
Thermo Scientific Chemicals
Dimethyltin dichloride, 95%
CAS: 753-73-1 | C2H6Cl2Sn | 219.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number FSA347820250
Price (MYR)
678.00
BOT
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
678.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDimethyltin dichloride
CAS753-73-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Suspected of damaging the unborn child.
Causes damage to organs through prolonged or repeated
Health Hazard 3GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.