3-Mercapto-1-hexanol, 96%
3-Mercapto-1-hexanol, 96%
3-Mercapto-1-hexanol, 96%
3-Mercapto-1-hexanol, 96%
Thermo Scientific Chemicals
3-Mercapto-1-hexanol, 96%
CAS: 51755-83-0 | C6H14OS | 134.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number FSA347560250
Price (MYR)
2,033.00
BOT
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,033.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Mercapto-1-hexanol
CAS51755-83-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0