4-Bromo-2-fluorobenzoyl chloride, 99%
4-Bromo-2-fluorobenzoyl chloride, 99%
4-Bromo-2-fluorobenzoyl chloride, 99%
4-Bromo-2-fluorobenzoyl chloride, 99%
Thermo Scientific Chemicals
4-Bromo-2-fluorobenzoyl chloride, 99%
CAS: 151982-51-3 | C7H3BrClFO | 237.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA347460250
Price (MYR)
1,662.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,662.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Bromo-2-fluorobenzoyl chloride
Name Note99%
CAS151982-51-3
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger