N-Acetyl-L-leucine, 99%
N-Acetyl-L-leucine, 99%
N-Acetyl-L-leucine, 99%
N-Acetyl-L-leucine, 99%
Thermo Scientific Chemicals
N-Acetyl-L-leucine, 99%
CAS: 1188-21-2 | C8H15NO3 | 173.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA347010050
Price (MYR)
367.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
367.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN-Acetyl-L-leucine
Name Note99%
CAS1188-21-2
CAS Min %98.5
CAS Max %100.0