D-Threonine, 98%
D-Threonine, 98%
D-Threonine, 98%
Thermo Scientific Chemicals
D-Threonine, 98%
CAS: 632-20-2 | C4H9NO3 | 119.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass Bottle
Catalog number FSA346970050
Price (MYR)
413.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
413.00
BOT
Add to cart
Specifications
Chemical Name or MaterialD-Threonine
Ammonium200ppm max.
CAS632-20-2
Arsenic (As)1ppm max.