D(-)-Phenylglycinamide, 99%
D(-)-Phenylglycinamide, 99%
D(-)-Phenylglycinamide, 99%
D(-)-Phenylglycinamide, 99%
Thermo Scientific Chemicals
D(-)-Phenylglycinamide, 99%
CAS: 6485-67-2 | C8H10N2O | 150.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA346700050
Price (MYR)
733.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
733.00
BOT
Add to cart
Specifications
Chemical Name or MaterialD(-)-Phenylglycinamide
Name Note99%
CAS6485-67-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement