Carminic acid, 70-90%
Carminic acid, 70-90%
Carminic acid, 70-90%
Carminic acid, 70-90%
Thermo Scientific Chemicals
Carminic acid, 70-90%
CAS: 1260-17-9 | C22H20O13 | 492.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA346630050
Price (MYR)
1,578.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,578.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCarminic acid
Name Note70-90%
CAS1260-17-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement: