1,2,3-1H-Triazole, 97%
1,2,3-1H-Triazole, 97%
1,2,3-1H-Triazole, 97%
1,2,3-1H-Triazole, 97%
Thermo Scientific Chemicals
1,2,3-1H-Triazole, 97%
CAS: 288-36-8 | C2H3N3 | 69.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA345480050
Price (MYR)
651.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
651.00
EA
Add to cart
Specifications
Chemical Name or Material1, 2, 3-1H-Triazole
CAS288-36-8
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0