Water, Extra Pure, Deionized
Water, Extra Pure, Deionized
Water, Extra Pure, Deionized
Water, Extra Pure, Deionized
Thermo Scientific Chemicals
Water, Extra Pure, Deionized
CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
5 L
Packaging:
Plastic Drum
Catalog number FSA345470250
Price (MYR)
1,225.00
EA
Add to cart
Quantity:
25 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (MYR)
1,225.00
EA
Add to cart
Specifications
Chemical Name or MaterialWater
CAS7732-18-5
ColorColorless
Health Hazard 1
Boiling Point100.0°C
View more
Biological buffer
RUO – Research Use Only