Methylcyclopentane, 95%
Methylcyclopentane, 95%
Methylcyclopentane, 95%
Methylcyclopentane, 95%
Thermo Scientific Chemicals
Methylcyclopentane, 95%
CAS: 96-37-7 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
Glass bottle
Catalog number FSA345245000
Price (MYR)
963.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
963.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethylcyclopentane
Name Note95%
CAS96-37-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.