Iron(III) sulfate pentahydrate, 97%
Iron(III) sulfate pentahydrate, 97%
Iron(III) sulfate pentahydrate, 97%
Iron(III) sulfate pentahydrate, 97%
Thermo Scientific Chemicals
Iron(III) sulfate pentahydrate, 97%
CAS: 142906-29-4 | Fe2O12S3·5H2O | 489.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA345235000
Price (MYR)
583.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
583.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIron(III) sulfate pentahydrate
CAS142906-29-4
ColorBeige-Yellow to White
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May be corrosive to metals.