N,N-Dimethylglycine, 97%
N,N-Dimethylglycine, 97%
N,N-Dimethylglycine, 97%
N,N-Dimethylglycine, 97%
Thermo Scientific Chemicals
N,N-Dimethylglycine, 97%
CAS: 1118-68-9 | C4H9NO2 | 103.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA345200250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialN, N-Dimethylglycine
Name Note97%
CAS1118-68-9
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning