N,N'-Bis(2-hydroxyethyl)ethylenediamine, 95%
N,N'-Bis(2-hydroxyethyl)ethylenediamine, 95%
N,N'-Bis(2-hydroxyethyl)ethylenediamine, 95%
N,N'-Bis(2-hydroxyethyl)ethylenediamine, 95%
Thermo Scientific Chemicals
N,N'-Bis(2-hydroxyethyl)ethylenediamine, 95%
CAS: 4439-20-7 | C6H16N2O2 | 148.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA345180100
Price (MYR)
399.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
399.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN, N'-Bis(2-hydroxyethyl)ethylenediamine
CAS4439-20-7
CAS Min %94.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0