Aluminum phosphate, pure
Aluminum phosphate, pure
Aluminum phosphate, pure
Aluminum phosphate, pure
Thermo Scientific Chemicals
Aluminum phosphate, pure
CAS: 7784-30-7 | AlO4P | 121.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass Bottle
Catalog number FSA344445000
Price (MYR)
955.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
955.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAluminum phosphate
Melting Point>1500.0°C
CAS7784-30-7
ColorWhite
Physical FormPowder