Triethylenetetramine, 60%
Triethylenetetramine, 60%
Triethylenetetramine, 60%
Triethylenetetramine, 60%
Thermo Scientific Chemicals
Triethylenetetramine, 60%
CAS: 112-24-3 | C6H18N4 | 146.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
50 g
Catalog number FSA344075000
Price (MYR)
321.00
BOT
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
321.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTriethylenetetramine
Name Note60%
CAS112-24-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful in contact with skin.
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.