L(-)-Glutathione, oxidized hydrate, 95%
L(-)-Glutathione, oxidized hydrate, 95%
L(-)-Glutathione, oxidized hydrate, 95%
L(-)-Glutathione, oxidized hydrate, 95%
Thermo Scientific Chemicals
L(-)-Glutathione, oxidized hydrate, 95%
CAS: 27025-41-8 | C20H32N6O12S2 | 612.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA343840050
Price (MYR)
1,287.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,287.00
BOT
Add to cart
Specifications
Chemical Name or MaterialL(-)-Glutathione, oxidized hydrate
CAS27025-41-8
Health Hazard 1Warning
Health Hazard 2GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.