Sodium phosphate dibasic dihydrate, 99+%, for HPLC
Sodium phosphate dibasic dihydrate, 99+%, for HPLC
Sodium phosphate dibasic dihydrate, 99+%, for HPLC
Sodium phosphate dibasic dihydrate, 99+%, for HPLC
Thermo Scientific Chemicals
Sodium phosphate dibasic dihydrate, 99+%, for HPLC
CAS: 10028-24-7 | H5Na2O6P | 177.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA343815000
Price (MYR)
903.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
903.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium phosphate dibasic dihydrate
Melting Point243.0°C to 245.0°C
Name NoteFor HPLC
CAS10028-24-7