Thiophene, 99.5%, extra pure, benzene free
Thiophene, 99.5%, extra pure, benzene free
Thiophene, 99.5%, extra pure, benzene free
Thiophene, 99.5%, extra pure, benzene free
Thermo Scientific Chemicals
Thiophene, 99.5%, extra pure, benzene free
CAS: 110-02-1 | C4H4S | 84.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA342595000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialThiophene
Name Note99.5%
CAS110-02-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapour.
Harmful if swallowed.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.