4-Iodobenzyl bromide, 97%
4-Iodobenzyl bromide, 97%
4-Iodobenzyl bromide, 97%
4-Iodobenzyl bromide, 97%
Thermo Scientific Chemicals
4-Iodobenzyl bromide, 97%
CAS: 16004-15-2 | C7H6BrI | 296.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA342460250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Iodobenzyl bromide
CAS16004-15-2
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0