Sodium perchlorate, 98%, extra pure
Sodium perchlorate, 98%, extra pure
Sodium perchlorate, 98%, extra pure
Sodium perchlorate, 98%, extra pure
Thermo Scientific Chemicals
Sodium perchlorate, 98%, extra pure
CAS: 7601-89-0 | ClNaO4 | 122.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA342182500
Price (MYR)
636.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
636.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSodium perchlorate
Melting Point482.0°C
CAS7601-89-0
ColorColorless to Yellow
Health Hazard 1GHS Signal Word: Danger
View more
Oxidation reagent
RUO – Research Use Only