Patent Blue V, sodium salt
Patent Blue V, sodium salt
Patent Blue V, sodium salt
Patent Blue V, sodium salt
Thermo Scientific Chemicals
Patent Blue V, sodium salt
CAS: 20262-76-4 | C27H31N2NaO7S2 | 582.662 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA339331000
Price (MYR)
828.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
828.00
EA
Add to cart
Specifications
Chemical Name or MaterialPatent blue V, sodium salt
CAS20262-76-4
Health Hazard 2GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Dye Content80% min.