Methyl Red hydrochloride, ACS reagent
Methyl Red hydrochloride, ACS reagent
Methyl Red hydrochloride, ACS reagent
Methyl Red hydrochloride, ACS reagent
Thermo Scientific Chemicals
Methyl Red hydrochloride, ACS reagent
CAS: 63451-28-5 | C15H16ClN3O2 | 305.762 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA339321000
Price (MYR)
546.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
546.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethyl Red hydrochloride
CAS63451-28-5
EINECS Number264-190-9
ClarityPasses Test
Molecular FormulaC15H16ClN3O2