Sodium amide, 99%, extra pure
Sodium amide, 99%, extra pure
Sodium amide, 99%, extra pure
Sodium amide, 99%, extra pure
Thermo Scientific Chemicals
Sodium amide, 99%, extra pure
CAS: 7782-92-5 | H2NNa | 39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA339241000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Boiling Point400.0°C
Chemical Name or MaterialSodium amide
Melting Point210.0°C
Name Note99%
CAS7782-92-5