6-α-Methylprednisolone
6-α-Methylprednisolone
6-α-Methylprednisolone
6-α-Methylprednisolone
Thermo Scientific Chemicals
6-α-Methylprednisolone
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA338800010
Price (MYR)
565.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
565.00
EA
Add to cart
Specifications
Chemical Name or Material6-α-Methylprednisolone
CAS83-43-2
Health Hazard 1GHS Signal Word: Danger
Melting Point227.0°C to 231.0°C
EINECS Number201-476-4