3-Methoxy-1,2-propanediol, 98%
3-Methoxy-1,2-propanediol, 98%
3-Methoxy-1,2-propanediol, 98%
3-Methoxy-1,2-propanediol, 98%
Thermo Scientific Chemicals
3-Methoxy-1,2-propanediol, 98%
CAS: 623-39-2 | C4H10O3 | 106.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA338390050
Price (MYR)
514.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
514.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Methoxy-1, 2-propanediol
CAS623-39-2
CAS Min %97.5
CAS Max %100.0