Check Order Status
4'-Hydroxy-4-biphenylcarbonitrile, 95%, Thermo Scientific™
Element Search
Structure Search
Thermo Scientific Chemicals
4'-Hydroxy-4-biphenylcarbonitrile, 95%, Thermo Scientific™
Have Questions?
Change view
Quantity:
1 g
Packaging:
Glass bottle
Catalog number
FSA338340010
Price (MYR)
223.00
BOT
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
223.00
BOT
Add to cart
Specifications
Chemical Name or Material
4′-Hydroxy-4-biphenylcarbonitrile
Name Note
95%
CAS
19812-93-2
CAS Min %
94.0
Health Hazard 1
GHS Signal Word:
Warning