Lithium bis(trimethylsilyl)amide, 95%
Lithium bis(trimethylsilyl)amide, 95%
Lithium bis(trimethylsilyl)amide, 95%
Lithium bis(trimethylsilyl)amide, 95%
Thermo Scientific Chemicals
Lithium bis(trimethylsilyl)amide, 95%
CAS: 4039-32-1 | C6H18LiNSi2 | 167.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Packaging:
Glass bottle
Catalog number FSA338142500
Price (MYR)
4,386.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,386.00
EA
Add to cart
Specifications
Chemical Name or MaterialLithium bis(trimethylsilyl)amide
CAS4039-32-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable solid.
Causes severe skin burns and eye damage.
Reacts violently with water.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse