n-Octyltriethoxysilane, 97%
n-Octyltriethoxysilane, 97%
n-Octyltriethoxysilane, 97%
n-Octyltriethoxysilane, 97%
Thermo Scientific Chemicals
n-Octyltriethoxysilane, 97%
CAS: 2943-75-1 | C14H32O3Si | 276.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA338081000
Price (MYR)
540.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
540.00
BOT
Add to cart
Specifications
Chemical Name or Materialn-Octyltriethoxysilane
Name Note97%
CAS2943-75-1
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning