4-Methylumbelliferyl-beta-D-glucuronide dihydrate, 98%
4-Methylumbelliferyl-beta-D-glucuronide dihydrate, 98%
4-Methylumbelliferyl-beta-D-glucuronide dihydrate, 98%
4-Methylumbelliferyl-beta-D-glucuronide dihydrate, 98%
Thermo Scientific Chemicals
4-Methylumbelliferyl-beta-D-glucuronide dihydrate, 98%
CAS: 6160-80-1 | C16H16O9·2H2O | 388.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number FSA337220050
Price (MYR)
1,073.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,073.00
EA
Add to cart
Specifications
Chemical Name or Material4-Methylumbelliferyl-β-D-glucuronide dihydrate
CAS6160-80-1
Melting Point102.0°C
EINECS Number228-186-
Infrared SpectrumAuthentic