trans-Zeatin-riboside, 97%
trans-Zeatin-riboside, 97%
trans-Zeatin-riboside, 97%
trans-Zeatin-riboside, 97%
Thermo Scientific Chemicals
trans-Zeatin-riboside, 97%
CAS: 6025-53-2 | C15H21N5O5 | 351.36 g/mol
Have Questions?
Catalog NumberQuantity
FSA33720050050 mg
Catalog number FSA337200500
Price (MYR)
2,551.00
BOT
Add to cart
Quantity:
50 mg
Request bulk or custom format
Price (MYR)
2,551.00
BOT
Add to cart
Specifications
Chemical Name or Materialtrans-Zeatin-riboside
CAS6025-53-2
Health Hazard 1MOISTURE SENSITIVE
Melting Point179°C to 180°C