4-Methylumbelliferyl-beta-D-glucopyranoside, 99%
4-Methylumbelliferyl-beta-D-glucopyranoside, 99%
4-Methylumbelliferyl-beta-D-glucopyranoside, 99%
4-Methylumbelliferyl-beta-D-glucopyranoside, 99%
Thermo Scientific Chemicals
4-Methylumbelliferyl-beta-D-glucopyranoside, 99%
CAS: 18997-57-4 | C16H18O8 | 338.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
Packaging:
Glass bottle
Catalog number FSA337025000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Methylumbelliferyl-β-D-glucopyranoside
Name Note99%
CAS18997-57-4
CAS Min %98.5
Health Hazard 1MOISTURE SENSITIVE