3-Aminopropyldimethylmethoxysilane, 97%
3-Aminopropyldimethylmethoxysilane, 97%
3-Aminopropyldimethylmethoxysilane, 97%
3-Aminopropyldimethylmethoxysilane, 97%
Thermo Scientific Chemicals
3-Aminopropyldimethylmethoxysilane, 97%
CAS: 31024-26-7 | C6H17NOSi | 147.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA336600050
Price (MYR)
2,133.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,133.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Aminopropyldimethylmethoxysilane
Name Note97%
CAS31024-26-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger