3-Nitrophenyl isocyanate, 97%, Thermo Scientific Chemicals
3-Nitrophenyl isocyanate, 97%, Thermo Scientific Chemicals
3-Nitrophenyl isocyanate, 97%, Thermo Scientific Chemicals
3-Nitrophenyl isocyanate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Nitrophenyl isocyanate, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA336180250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-Nitrophenyl isocyanate
Name Note97%
CAS3320-87-4
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning