1,2-Epoxyhexane, 97%
1,2-Epoxyhexane, 97%
1,2-Epoxyhexane, 97%
1,2-Epoxyhexane, 97%
Thermo Scientific Chemicals
1,2-Epoxyhexane, 97%
CAS: 1436-34-6 | C6H12O | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number FSA335550250
Price (MYR)
456.00
BOT
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
456.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 2-Epoxyhexane
CAS1436-34-6
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0