Dowex™ 50WX8, 50-100 mesh, ion-exchange resin
Dowex™ 50WX8, 50-100 mesh, ion-exchange resin
Dowex™ 50WX8, 50-100 mesh, ion-exchange resin
Dowex™ 50WX8, 50-100 mesh, ion-exchange resin
Dowex™ 50WX8, 50-100 mesh, ion-exchange resin
CAS: 11119-67-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic Bottle
Catalog number FSA335335000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDowex™ 50WX8
Physical FormMoist Spheres
CAS11119-67-8
Particle Size15% max. (on 35 mesh, cumulative), 20% max. (through 80 mesh)
PackagingPlastic Bottle