1,2-Benzenedithiol, 96%
1,2-Benzenedithiol, 96%
1,2-Benzenedithiol, 96%
1,2-Benzenedithiol, 96%
Thermo Scientific Chemicals
1,2-Benzenedithiol, 96%
CAS: 17534-15-5 | C6H6S2 | 142.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA334920010
Price (MYR)
919.00
BOT
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
919.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 2-Benzenedithiol
CAS17534-15-5
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0