Docosanoic acid, 85%, technical
Docosanoic acid, 85%, technical
Docosanoic acid, 85%, technical
Docosanoic acid, 85%, technical
Thermo Scientific Chemicals
Docosanoic acid, 85%, technical
CAS: 112-85-6 | C22H44O2 | 340.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
View all
Glass bottle
Catalog number FSA334812500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDocosanoic acid
CAS112-85-6
Melting Point80.0°C to 82.0°C
ColorWhite to Yellow
EINECS Number204-010-8