(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%
(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%
(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%
(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%
Thermo Scientific Chemicals
(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%
CAS: 2964-48-9 | C9H12N2O4 | 212.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA334800250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material(1S, 2S)-2-Amino-1-(4-nitrophenyl)propane-1, 3-diol
CAS2964-48-9
CAS Min %95.0
CAS Max %100.0
Melting Point160°C to 166°C