Sodium caprylate, 98%
Sodium caprylate, 98%
Sodium caprylate, 98%
Sodium caprylate, 98%
Sodium caprylate, 98%
Thermo Scientific Chemicals
Sodium caprylate, 98%
CAS: 1984-06-1 | C8H15NaO2 | 166.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number FSA334601000
Price (MYR)
327.00
BOT
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
327.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSodium caprylate
Name Note98%
CAS1984-06-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: