2-Naphthalenethiol, 98+%
2-Naphthalenethiol, 98+%
2-Naphthalenethiol, 98+%
2-Naphthalenethiol, 98+%
Thermo Scientific Chemicals
2-Naphthalenethiol, 98+%
CAS: 91-60-1 | C10H8S | 160.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA334520050
Price (MYR)
419.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
419.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Naphthalenethiol
Name Note99%
CAS91-60-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning