Check Order Status
(2-Pyrimidylthio)acetic acid, 98%, Thermo Scientific™
Element Search
Structure Search
Thermo Scientific Chemicals
(2-Pyrimidylthio)acetic acid, 98%, Thermo Scientific™
Have Questions?
Change view
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number
FSA334490250
Price (MYR)
735.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
735.00
BOT
Add to cart
Specifications
Chemical Name or Material
(2-Pyrimidylthio)acetic acid
Name Note
98%
CAS
88768-45-0
CAS Min %
97.5
Health Hazard 1
GHS Signal Word:
Warning