Piperonyl butoxide, 90%, Tech.
Piperonyl butoxide, 90%, Tech.
Piperonyl butoxide, 90%, Tech.
Piperonyl butoxide, 90%, Tech.
Thermo Scientific Chemicals
Piperonyl butoxide, 90%, Tech.
CAS: 51-03-6 | C19H30O5 | 338.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA334161000
Price (MYR)
368.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
368.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPiperonyl butoxide
Name Notetech., 90%
CAS51-03-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Very toxic to aquatic life with long lasting effects.