6-Methoxy-1-indanone, 98%
6-Methoxy-1-indanone, 98%
6-Methoxy-1-indanone, 98%
6-Methoxy-1-indanone, 98%
Thermo Scientific Chemicals
6-Methoxy-1-indanone, 98%
CAS: 13623-25-1 | C10H10O2 | 162.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
View all
Glass Bottle
Catalog number FSA334070250
Price (MYR)
2,005.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,005.00
EA
Add to cart
Specifications
Chemical Name or Material6-Methoxy-1-indanone
CAS13623-25-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER