Carboxymethyl cellulose, sodium salt, average M.W. 700000 (DS=0.9)
Carboxymethyl cellulose, sodium salt, average M.W. 700000 (DS=0.9)
Carboxymethyl cellulose, sodium salt, average M.W. 700000 (DS=0.9)
Carboxymethyl cellulose, sodium salt, average M.W. 700000 (DS=0.9)
Carboxymethyl cellulose, sodium salt, average M.W. 700000 (DS=0.9)
Thermo Scientific Chemicals
Carboxymethyl cellulose, sodium salt, average M.W. 700000 (DS=0.9)
CAS: 9004-32-4 | (C12 H14 O9 R6)n | 263.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Packaging:
View all
Glass bottle
Catalog number FSA332641000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCarboxymethylecellulose, sodium salt
ColorWhite to Beige
Name NoteAverage M.W. 700000 (DS=0.9)
pH6.5 to 8 (1% soln.)
CAS9004-32-4
View more
Suitable for battery materials development.
RUO – Research Use Only