D-erythro-Sphingosine, 99+%, synthetical
D-erythro-Sphingosine, 99+%, synthetical
D-erythro-Sphingosine, 99+%, synthetical
D-erythro-Sphingosine, 99+%, synthetical
Thermo Scientific Chemicals
D-erythro-Sphingosine, 99+%, synthetical
CAS: 123-78-4 | C18H37NO2 | 299.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
Packaging:
Glass bottle
Catalog number FSA331532500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialD-erythro-Sphingosine
Name Note99+%
CAS123-78-4
Melting Point81.0°C to 82.0°C
ColorWhite to Cream