DL-6-Methoxy-alpha-methyl-2-naphthalenemethanol, 98%
DL-6-Methoxy-alpha-methyl-2-naphthalenemethanol, 98%
DL-6-Methoxy-alpha-methyl-2-naphthalenemethanol, 98%
DL-6-Methoxy-alpha-methyl-2-naphthalenemethanol, 98%
Thermo Scientific Chemicals
DL-6-Methoxy-alpha-methyl-2-naphthalenemethanol, 98%
CAS: 77301-42-9 | C13H14O2 | 202.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA331500050
Price (MYR)
698.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
698.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDL-6-Methoxy-α-methyl-2-naphthalenemethanol
Name Note98%
CAS77301-42-9
Melting Point111.0°C to 114.0°C
ColorBeige-Yellow to White