2'-Hydroxy-5'-methoxyacetophenone, 99%
2'-Hydroxy-5'-methoxyacetophenone, 99%
2'-Hydroxy-5'-methoxyacetophenone, 99%
2'-Hydroxy-5'-methoxyacetophenone, 99%
Thermo Scientific Chemicals
2'-Hydroxy-5'-methoxyacetophenone, 99%
CAS: 705-15-7 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA331240050
Price (MYR)
412.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
412.00
BOT
Add to cart
Specifications
Chemical Name or Material2'-Hydroxy-5'-methoxyacetophenone
Name Note99%
CAS705-15-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning