2,3-Dichloroquinoxaline, 97%
2,3-Dichloroquinoxaline, 97%
2,3-Dichloroquinoxaline, 97%
2,3-Dichloroquinoxaline, 97%
Thermo Scientific Chemicals
2,3-Dichloroquinoxaline, 97%
CAS: 2213-63-0 | C8H4Cl2N2 | 199.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA330830100
Price (MYR)
579.00
EA
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
579.00
EA
Add to cart
Specifications
Chemical Name or Material2, 3-Dichloroquinoxaline
Name Note97%
CAS2213-63-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger